63020 Fluka

Magnesium chloride solution

volumetric, 1.0 M MgCl2

DOWNLOAD MSDS (PDF)
  • CAS Number 7786-30-3

  • Linear Formula MgCl2

  • Molecular Weight 95.21

  •  PubChem Substance ID 24882158

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, G - M, Salt Solutions, Titration,
quality   volumetric
shelf life   (limited shelf life, expiry date on the label)
concentration   1.0 M MgCl2

Description

General description

Visit our Titration Center to learn more.

Price and Availability

Customers Also Viewed

Calcium chloride solution

volumetric, 1.0 M CaCl2

Magnesium chloride

anhydrous, ≥98%

for molecular biology, 1.00 M±0.01 M

PCR Reagent, 25 mM MgCI2 solution for PCR

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?