EMAIL THIS PAGE TO A FRIEND

63063 Fluka

Magnesium chloride

purum, anhydrous, ≥98.0% (calc. based on dry substance, KT)

Synonym: Magnogene

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Magnesium, Synthetic Reagents
grade   purum
assay   ≥98.0% (calc. based on dry substance, KT)
impurities   ≤0.5% insoluble matter in water
loss   ≤3% loss on drying
mp   714 °C(lit.)
density   2.32 g/mL at 25 °C(lit.)

Description

Caution

may contain particles

Other Notes

Eliminates non-specific staining in the alcian blue staining of proteoglycans in polyacrylamide gels1

This product has been replaced by M8266-SIGMA | Magnesium chloride anhydrous, ≥98%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Additional Products to Explore

Calcium chloride

anhydrous, granular, ≤7.0 mm, ≥93.0%

Magnesium chloride hexahydrate

ACS reagent, 99.0-102.0%

Magnesium sulfate

anhydrous, ReagentPlus®, ≥99.5%

Potassium chloride

BioXtra, ≥99.0%

Sodium chloride

BioXtra, ≥99.5% (AT)

Customers Also Viewed

Magnesium chloride

anhydrous, beads, −10 mesh, 99.99% trace metals basis

Magnesium chloride

anhydrous, beads, −10 mesh, 99.9% trace metals basis

Magnesium chloride

powder, <200 μm

Magnesium chloride hexahydrate

BioXtra, ≥99.0%

Magnesium chloride hexahydrate

ACS reagent, 99.0-102.0%

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OM2800000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. Alcian blue staining of proteoglycans in polyacrylamide gels using the "critical electrolyte concentration" approach. R.S. Wall, T.J. Gyi Anal. Biochem. 175, 298, (1988)

Loading...


Direct and real-time observation of rotary movement of a DNA nanomechanical device. Rajendran A, Endo M, Hidaka K, et al. J. Am. Chem. Soc. 135(3), 1117-23, (2013)

Loading...


Effects of proteoliposome composition and draw solution types on separation performance of aquaporin-based proteoliposomes: implications for seawater desalination using aquaporin-based biomimetic membranes. Zhao Y, Vararattanavech A, Li X, et al. Environ. Sci. Technol. 47(3), 1496-503, (2013)

Loading...


Directly probing the effects of ions on hydration forces at interfaces. Kilpatrick JI, Loh SH, and Jarvis SP J. Am. Chem. Soc. 135(7), 2628-34, (2013)

Loading...


The nucleoid-associated protein Dan organizes chromosomal DNA through rigid nucleoprotein filament formation in E. coli during anoxia. Lim CJ, Lee SY, Teramoto J, et al. Nucleic Acids Res. 41(2), 746-53, (2013)

Loading...


Cognitive changes after saline or plasmalyte infusion in healthy volunteers: a multiple blinded, randomized, cross-over trial. Story DA, Lees L, Weinberg L, et al. Anesthesiology 119(3), 569-75, (2013)

Loading...


Divalent cations enhance fluoride binding to Streptococcus mutans and Streptococcus sanguinis cells and subsequently inhibit bacterial acid production. Domon-Tawaraya H, Nakajo K, Washio J, et al. Caries Res. 47(2), 141-9, (2013)

Loading...


Acclimation of leaves to low light produces large grana: the origin of the predominant attractive force at work. Jia H, Liggins JR, and Chow WS Philos. Trans. R. Soc. Lond., B, Biol. Sci. 367(1608), 3494-502, (2012)

Loading...


A hint on pharmacological and toxicological differences between magnesium chloride and magnesium sulphate, or of scallops and men. Durlach J, Bara M, and Theophanides T Magnes. Res. 9(3), 217-9, (1996)

Loading...


A protocol for the 0.9% saline versus Plasma-Lyte 148 for intensive care fluid therapy (SPLIT) study. Reddy SK, Bailey MJ, Beasley RW, et al. Crit. Care Resusc. 16(4), 274-9, (2014)

Loading...


[The effectiveness of certain magnesium salts in nephrolithiasis caused by the use of sodium oxalate and celecoxib]. Spasov AA, Iezhitsa IN, Kharitonova MV, et al. Urologiia. (1), 29-34, (2013)

Loading...


Purification, optimization of assay, and stability studies of dextransucrase isolated from Weissella cibaria JAG8. Mohan Rao TJ and Goyal A Prep Biochem Biotechnol. 43(4), 329-41, (2013)

Loading...


Rapid and efficient synthesis of nucleoside polyphosphates and their conjugates using sulfonyl imidazolium salts. Mohamady S and Taylor SD Curr. Protoc. Nucleic Acid Chem. Chapter 13, Unit13.11, (2012)

Loading...


Observing a DNA polymerase choose right from wrong. Freudenthal BD, Beard WA, Shock DD, et al. Cell 154(1), 157-68, (2013)

Loading...


Concentration polarization phenomenon during the nanofiltration of multi-ionic solutions: influence of the filtrated solution and operating conditions. Déon S, Dutournié P, Fievet P, et al. Water Res. 47(7), 2260-72, (2013)

Loading...


Cell specific aptamer-photosensitizer conjugates as a molecular tool in photodynamic therapy. Prabodhika Mallikaratchy et al ChemMedChem 3, 425-8, (2010)

Loading...


Inflammatory bowel disease: MR- and SPECT/CT-based macrophage imaging for monitoring and evaluating disease activity in experimental mouse model--pilot study. Wu Y, Briley-Saebo K, Xie J, et al. Radiology 271(2), 400-7, (2014)

Loading...


Cation-dependent folding of 3' cap-independent translation elements facilitates interaction of a 17-nucleotide conserved sequence with eIF4G. Kraft JJ, Treder K, Peterson MS, et al. Nucleic Acids Res. 41(5), 3398-413, (2013)

Loading...


Characterising protein, salt and water interactions with combined vibrational spectroscopic techniques. Perisic N, Afseth NK, Ofstad R, et al. Food Chem. 138(1), 679-86, (2013)

Loading...


Manganese accumulation in bone following chronic exposure in rats: steady-state concentration and half-life in bone. O'Neal SL, Hong L, Fu S, et al. Toxicol. Lett. 229(1), 93-100, (2014)

Loading...


Squid have nociceptors that display widespread long-term sensitization and spontaneous activity after bodily injury. Crook RJ, Hanlon RT, and Walters ET J. Neurosci. 33(24), 10021-6, (2013)

Loading...


Investigation into the controversial association of Streptococcus gallolyticus with colorectal cancer and adenoma. Abdulamir AS BMC Cancer 9, 403, (2009)

Loading...


Salts drive controllable multilayered upright assembly of amyloid-like peptides at mica/water interface. Dai B, Kang SG, Huynh T, et al. Proc. Natl. Acad. Sci. U. S. A. 110(21), 8543-8, (2013)

Loading...


Magnesium chloride or magnesium sulfate: a genuine question. Durlach J, Guiet-Bara A, Pagès N, et al. Magnes. Res. 18(3), 187-92, (2005)

Loading...


Halobellus inordinatus sp. nov., from a marine solar saltern and an inland salt lake of China. Qiu XX, Mou YZ, Zhao ML, et al. Int. J. Syst. Evol. Microbiol. 63(Pt 11), 3975-80, (2013)

Loading...


Effects of soybean protein composition and processing conditions on silken tofu properties. Yang A and James AT J. Sci. Food Agric. 93(12), 3065-71, (2013)

Loading...


Importance of magnesium chloride repletion after myocardial infarction. Sheehan J Am. J. Cardiol. 63(14), 35G-38G, (1989)

Loading...


Effects of NH4Cl and MgCl(2) on pretreatment and xylan hydrolysis of miscanthus straw. Kang KE, Park DH, and Jeong GT Carbohydr. Polym. 92(2), 1321-6, (2013)

Loading...


K(+) , Na(+) , and Mg(2+) on DNA translocation in silicon nitride nanopores. Uplinger J, Thomas B, Rollings R, et al. Electrophoresis 33(23), 3448-57, (2012)

Loading...


Oral magnesium supplementation improves the metabolic profile of metabolically obese, normal-weight individuals: a randomized double-blind placebo-controlled trial. Rodríguez-Moran M and Guerrero-Romero F Arch. Med. Res. 45(5), 388-93, (2014)

Loading...


Leishmania amazonensis: characterization of an ecto-pyrophosphatase activity. Freitas-Mesquita AL, Fonseca-de-Souza AL, and Meyer-Fernandes JR Exp. Parasitol. 137, 8-13, (2014)

Loading...


Increased frequency and severity of developmental deformities in rough-skinned newt (Taricha granulosa) embryos exposed to road deicing salts (NaCl & MgCl2). Hopkins GR, French SS, and Brodie ED Environ. Pollut. 173, 264-9, (2013)

Loading...


Maternal magnesium supplementation reduces intrauterine growth restriction and suppresses inflammation in a rat model. Roman A, Desai N, Rochelson B, et al. Am. J. Obstet. Gynecol. 208(5), 383.e1-7, (2013)

Loading...


Bacterial maximum non-inhibitory and minimum inhibitory concentrations of different water activity depressing solutes. Cebrián G, Arroyo C, Mañas P, et al. Int. J. Food Microbiol. 188, 67-74, (2014)

Loading...


Characterization of a variant of Xanthomonas citri subsp. citri that triggers a host-specific defense response. Chiesa MA, Siciliano MF, Ornella L, et al. Phytopathology 103(6), 555-64, (2013)

Loading...


Effect of salt reduction on growth of Listeria monocytogenes in meat and poultry systems. Harper NM and Getty KJ J. Food Sci. 77(12), M669-74, (2012)

Loading...


Differences in photosynthetic performance and its correlation with growth among tomato cultivars in response to different salts. Nebauer SG, Sánchez M, Martínez L, et al. Plant Physiol. Biochem. 63, 61-9, (2013)

Loading...


Common trace elements alleviate pain in an experimental mouse model. Tamba BI, Leon MM, and Petreus T J. Neurosci. Res. 91(4), 554-61, (2013)

Loading...


Structures of septin filaments prepared from rat brain and expressed in bacteria. Maimaitiyiming M, Kumanogoh H, Nakamura S, et al. Protein Expr. Purif. 87(2), 67-71, (2013)

Loading...


Meta-analysis of randomized controlled trials on magnesium in addition to beta-blocker for prevention of postoperative atrial arrhythmias after coronary artery bypass grafting. Wu X, Wang C, Zhu J, et al. BMC Cardiovasc. Disord. 13, 5, (2013)

Loading...


Elevated ferric, calcium and magnesium ions in the brain induce protein aggregation in brain mitochondria. Alleyne T, Mohan N, and Adogwa A West Indian Med. J. 61(2), 122-7, (2012)

Loading...


Magnesium supplementation reduces intrauterine growth restriction and inflammation: how and why? Das UN Am. J. Obstet. Gynecol. 209(5), 497-8, (2013)

Loading...


Reply: To PMID 23474429. Metz C, Roman A, Gupta M, et al. Am. J. Obstet. Gynecol. 209(5), 498-9, (2013)

Loading...


Heparin selectively affects the quantification of microRNAs in human blood samples. Boeckel JN, Thomé CE, Leistner D, et al. Clin. Chem. 59(7), 1125-7, (2013)

Loading...


Merck 14,5662

Corp MSDS 1 (2), 2178:C / RegBook 1 (3), 3321:A / Sax 6, 1722

Related Products

replaced by

Product #

Description

Add to Cart

M8266 Magnesium chloride, anhydrous, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?