63068 Fluka

Magnesium chloride hexahydrate

BioUltra, for molecular biology, ≥99.0% (KT)

DOWNLOAD MSDS (PDF)
  • CAS Number 7791-18-6

  • Linear Formula MgCl2 · 6H2O

  • Molecular Weight 203.30

  •  EC Number 232-094-6

  •  MDL number MFCD00149781

  •  PubChem Substance ID 24882331

Properties

Related Categories BioUltra, DNA & RNA Purification, Molecular Biology, Molecular Biology Reagents, Optimization Reagents,
grade   for molecular biology
product line   BioUltra
assay   ≥99.0% (KT)
impurities   DNases, none detected
  RNases, none detected
  insoluble matter, passes filter test
  phosphatases, none detected
  proteases, none detected
  ≤0.0002% total nitrogen (N)
pH   5.0-7.5 (25 °C, 1 M in H2O)
solubility   H2O: soluble1 M at 20 °C, clear, colorless
anion traces   phosphate (PO43-): ≤5 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤2000 mg/kg
  Li: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤3000 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: 0.030
  λ: 280 nm Amax: 0.025

Description

Caution

may agglomerate

Other Notes

Activation of restriction endonucleases1

Price and Availability

Customers Also Viewed

Calcium chloride dihydrate

for molecular biology, ≥99%

Magnesium chloride

anhydrous, ≥98%

Magnesium chloride hexahydrate

BioXtra, ≥99.0%

Magnesium chloride hexahydrate

BioReagent, suitable for cell culture, suitable for insect cell culture

Magnesium chloride hexahydrate

ACS reagent, 99.0-102.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?