EMAIL THIS PAGE TO A FRIEND

63107 Fluka

Magnesium oxide

tested according to Ph.Eur., heavy

Synonym: Magnesii oxidum ponderosum

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Magnesium, Materials Science, Metal and Ceramic Science,
pharmacopeia   testing conforms to Pharmacopeia
mp   2852 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Magnesium oxide

light (95%)

Magnesium oxide

BioUltra, ≥97.0% (calcined substance, KT)

Magnesium oxide

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)

Magnesium oxide

heavy, purum p.a., ≥98.0% (based on calcined substance, KT)

Magnesium oxide

tested according to Ph.Eur., light

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OM3850000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Determination of Water Content in Magnesium oxide Using Karl Fischer Titration

KF solutions react slightly acidic and can dissolve oxides, hydroxides and carbonates. This leads to formation of water which also gets titrated. Individual oxides or carbonates react, however, in di...
Keywords: Titrations

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Reply to MM Joosten et al. Veronese N, Manzato E, and Sergi G Am. J. Clin. Nutr. 101(1), 241, (2015)

Loading...


Assessing magnesium by 24-h urinary excretion. Joosten MM, Gansevoort RT, and Bakker SJ Am. J. Clin. Nutr. 101(1), 240-1, (2015)

Loading...


A prospective study on the clinical effectiveness of the stainless steel crown Herbst appliance. Latkauskiene D, Jakobsone G, and McNamara JA Prog. Orthod. 13(2), 100-8, (2012)

Loading...


[A case of the milk-alkali syndrome with a small amount of milk and magnesium oxide ingestion--the contribution of sustained metabolic alkalosis induced by hypertonic dehydration]. Yamada T, Nakanishi T, Uyama O, et al. Nihon Jinzo Gakkai Shi 33(6), 581-6, (1991)

Loading...


The entropies of adsorbed molecules. Campbell CT and Sellers JR J. Am. Chem. Soc. 134(43), 18109-15, (2012)

Loading...


Biocompatible nanostructured magnesium oxide-chitosan platform for genosensing application. Patel MK, Ali MA, Zafaryab M, et al. Biosens. Bioelectron. 45, 181-8, (2013)

Loading...


Transverse spin Seebeck effect versus anomalous and planar Nernst effects in Permalloy thin films. Schmid M, Srichandan S, Meier D, et al. Phys. Rev. Lett. 111(18), 187201, (2013)

Loading...


First-principles prediction of new photocatalyst materials with visible-light absorption and improved charge separation: surface modification of rutile TiO₂ with nanoclusters of MgO and Ga₂O₃. Nolan M ACS Appl. Mater. Interfaces 4(11), 5863-71, (2012)

Loading...


The structural changes of the bagasse hemicelluloses during the cooking process involving active oxygen and solid alkali. Shi JB, Yang QL, Lin L, et al. Carbohydr. Res. 359, 65-9, (2012)

Loading...


Atomic scale characterization of interfacial water near an oxide surface using molecular dynamics simulations. Deshmukh SA and Sankaranarayanan SK Phys. Chem. Chem. Phys. 14(44), 15593-605, (2012)

Loading...


A comparative synthesis and physicochemical characterizations of Ni/Al2O3-MgO nanocatalyst via sequential impregnation and sol-gel methods used for CO2 reforming of methane. Aghamohammadi S, Haghighi M, and Karimipour S J. Nanosci. Nanotechnol. 13(7), 4872-82, (2013)

Loading...


Hydrogenation of quinolines, alkenes, and biodiesel by palladium nanoparticles supported on magnesium oxide. Rahi R, Fang M, Ahmed A, et al. Dalton Trans. 41(48), 14490-7, (2012)

Loading...


Fabrication of polypyrrole/graphene oxide composite nanosheets and their applications for Cr(VI) removal in aqueous solution. Li S, Lu X, Xue Y, et al. PLoS ONE 7(8), e43328, (2012)

Loading...


Characterization of changes of lignin structure in the processes of cooking with solid alkali and different active oxygen. Yang Q, Shi J, Lin L, et al. Bioresour. Technol. 123, 49-54, (2012)

Loading...


Improved virus removal in ceramic depth filters modified with MgO. Michen B, Fritsch J, Aneziris C, et al. Environ. Sci. Technol. 47(3), 1526-33, (2013)

Loading...


Mechanical activation of CaO-based adsorbents for CO(2) capture. Sayyah M, Lu Y, Masel RI, et al. ChemSusChem 6(1), 193-8, (2013)

Loading...


Surface properties of calcium and magnesium oxide nanopowders grafted with unsaturated carboxylic acids studied with inverse gas chromatography. Maciejewska M, Krzywania-Kaliszewska A, and Zaborski M J. Chromatogr. A 1257, 141-8, (2012)

Loading...


Effect of partial crystallization on the mechanical properties and cytotoxicity of bioactive glass from the 3CaO.P(2)O(5)-SiO(2)-MgO system. Daguano JK, Strecker K, Ziemath EC, et al. J. Mech. Behav. Biomed. Mater. 14, 78-88, (2012)

Loading...


Using cathodoluminescence spectroscopy of cretaceous calcareous microfossils to distinguish biogenic from early-diagenetic calcite. Wendler JE, Wendler I, Rose T, et al. Microsc. Microanal. 18(6), 1313-21, (2012)

Loading...


Microstructure and mechanical properties of MgO-stabilized ZrO₂-Al₂O₃ dental composites. Abi CB, Emrullahoğlu OF, and Said G J. Mech. Behav. Biomed. Mater. 18, 123-31, (2013)

Loading...


KF-loaded mesoporous Mg-Fe bi-metal oxides: high performance transesterification catalysts for biodiesel production. Tao G, Hua Z, Gao Z, et al. Chem. Commun. (Camb.) 49(73), 8006-8, (2013)

Loading...


Nano-based antileishmanial agents: a toxicological study on nanoparticles for future treatment of cutaneous leishmaniasis. Jebali A and Kazemi B Toxicol. In Vitro 27(6), 1896-904, (2013)

Loading...


Micro and nano MgO particles for the improvement of fracture toughness of bone-cement interfaces. Khandaker M, Li Y, and Morris T J. Biomech. 46(5), 1035-9, (2013)

Loading...


Synthesis of mixed ceramic Mg(x)Zn(1-x)O nanofibers via Mg2+ doping using sol-gel electrospinning. Aykut Y, Parsons GN, Pourdeyhimi B, et al. Langmuir 29(12), 4159-66, (2013)

Loading...


Efficient enzymatic hydrolysis of the bagasse pulp prepared with active oxygen and MgO-based solid alkali. Xie T, Lin L, Pang C, et al. Carbohydr. Polym. 94(2), 807-13, (2013)

Loading...


In vivo corrosion mechanism by elemental interdiffusion of biodegradable Mg-Ca alloy. Jung JY, Kwon SJ, Han HS, et al. J. Biomed. Mater. Res. B. Appl. Biomater. 100(8), 2251-60, (2012)

Loading...


Isotope concentrations from 24-h urine and 3-h serum samples can be used to measure intestinal magnesium absorption in postmenopausal women. Hansen KE, Nabak AC, Johnson RE, et al. J. Nutr. 144(4), 533-7, (2014)

Loading...


Charge localization on the hexa-interstitial cluster in MgO. Mulroue J, Uberuaga BP, and Duffy DM J. Phys. Condens. Matter 25(6), 065502, (2013)

Loading...


Monte Carlo simulation and free energies of mixed oxide nanoparticles. Purton JA, Parker SC, and Allan NL Phys. Chem. Chem. Phys. 15(17), 6219-25, (2013)

Loading...


Acid mine drainage in the Iberian Pyrite Belt: 2. Lessons learned from recent passive remediation experiences. Ayora C, Caraballo MA, Macias F, et al. Environ. Sci. Pollut. Res. Int. 20(11), 7837-53, (2013)

Loading...


Nanostructured manganese oxide particles from coordination complex decomposition and their catalytic properties for ethanol oxidation. Palza H, Maturana A, Gracia F, et al. J. Nanosci. Nanotechnol. 12(10), 8087-93, (2012)

Loading...


Theoretical investigation of surface reactions of lactic acid on MgO clusters. Pandey LB and Aikens CM J. Phys. Chem. A 117(4), 765-70, (2013)

Loading...


Adherence of Streptococcus mutans to orthodontic band cements. Gonzalez-Perez JC, Scougall-Vilchis RJ, Contreras-Bulnes R, et al. Aust. Dent. J. 57(4), 464-9, (2012)

Loading...


Electro-physical properties of composites with nano-sized oxides. Lee SH and Choi Y J. Nanosci. Nanotechnol. 13(11), 7610-4, (2013)

Loading...


Growth mechanism of cubic MgO granule via common ion effect. Choi SB, Kim NW, Lee DK, et al. J. Nanosci. Nanotechnol. 13(11), 7577-80, (2013)

Loading...


Structural and optical properties of ZnO nanorods on Mg0.2Zn0.8O seed layers grown by hydrothermal method. Kim MS, Kim do Y, Kim SO, et al. J. Nanosci. Nanotechnol. 13(5), 3408-12, (2013)

Loading...


Sustain and address discharge characteristics of AC-PDP with MgO surface coated by MgO nano crystal powders. Kim JH, Park CS, Park HD, et al. J. Nanosci. Nanotechnol. 13(5), 3270-5, (2013)

Loading...


Droplet sampling of an oil-based and two water-based antievaporant ultra-low volume insecticide formulations using Teflon- and magnesium oxide-coated slides. Chaskopoulou A, Latham MD, Pereira RM, et al. J. Am. Mosq. Control Assoc. 29(2), 173-6, (2013)

Loading...


Kinetics of struvite precipitation in synthetic biologically treated swine wastewaters. Capdevielle A, Sýkorová E, Béline F, et al. Environ. Technol. 35(9-12), 1250-62, (2014)

Loading...


Full-mouth rehabilitation for a patient with dentinogenesis imperfecta: a clinical report. Bencharit S, Border MB, Mack CR, et al. J. Oral Implantol. 40(5), 593-600, (2014)

Loading...


Evaluation of mechanical properties of five cements for orthodontic band cementation. Aguiar DA, Ritter DE, Rocha R, et al. Braz. Oral Res. 27(2), 136-41, (2013)

Loading...


Magnesium oxide-pyridoxine therapy for recurrent urolithiasis. Nutr. Rev. 34(1), 18-20, (1976)

Loading...


Merck 14,5677

Fieser 1,633 / Fieser 2,256 / Fieser 7,221 / Fieser 16,200

Corp MSDS 1 (2), 2180:C / RegBook 1 (3), 3273:F / Sax 6, 1724

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?