63200 Fluka

Maleic anhydride

puriss., ≥99.0% (NT)

DOWNLOAD MSDS (PDF)

Synonym: 2,5-Furandione

  • CAS Number 108-31-6

  • Empirical Formula (Hill Notation) C4H2O3

  • Molecular Weight 98.06

  •  Beilstein Registry Number 106909

  •  EC Number 203-571-6

  •  MDL number MFCD00005518

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides, Chemical Synthesis, Organic Building Blocks More...
vapor density   3.4 (vs air)
vapor pressure   0.16 mmHg ( 20 °C)
grade   puriss.
assay   ≥99.0% (NT)
autoignition temp.   870 °F
expl. lim.   7.1 %
bp   200 °C(lit.)
mp   51-56 °C(lit.)
  52-54 °C

Description

Other Notes

For the complete dissociation of enzymes into soluble subunits1; For the reversible blocking of amino groups (lysine reagent); P.J.G. Butler, J.I. Harris, B.S. Hartley, R. Lebermann2; For the maleylation of proteins3

Price and Availability

Customers Also Viewed

Maleic anhydride

95% (may contain up to 5% maleic acid)

Maleic anhydride

briquettes, 99%

Maleic anhydride

powder, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?