63320 Fluka

Diethyl malonate

puriss., ≥99.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: Malonic acid diethyl ester

  • CAS Number 105-53-3

  • Linear Formula CH2(COOC2H5)2

  • Molecular Weight 160.17

  •  Beilstein Registry Number 774687

  •  EC Number 203-305-9

  •  MDL number MFCD00009195

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor density   5.52 (vs air)
vapor pressure   1 mmHg ( 40 °C)
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.413(lit.)
  n20/D 1.413
bp   199 °C(lit.)
mp   −51-−50 °C(lit.)
density   1.055 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Diethyl malonate

purum, ≥97.0% (GC)

Diethyl malonate

ReagentPlus®, 99%

Diethyl malonate

≥98%

Salicylaldehyde

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?