• USA Home
  • 63557 - Mannan from Saccharomyces cerevisiae

EMAIL THIS PAGE TO A FRIEND

63557 Fluka

Mannan from Saccharomyces cerevisiae

Purchase

Properties

optical activity   [α]/D +77±5°, c = 1 in H2O
storage temp.   2-8°C

Description

Preparation Note

by the Cetavlon method (S.A. Barker, et.al., Chem. Ind., S. 330 (1957))

Price and Availability

Customers Also Viewed

substrate for galactomannanase

Mannan from <I>Saccharomyces cerevisiae</I>

prepared by alkaline extraction

prepared by detergent extraction

saline suspension

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
OP1949500

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?