63660 Fluka

(−)-Menthol

analytical standard, for terpene analysis

DOWNLOAD MSDS (PDF)

Synonym: (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

  • CAS Number 2216-51-5

  • Empirical Formula (Hill Notation) C10H20O

  • Molecular Weight 156.27

  •  Beilstein Registry Number 1902293

  •  EC Number 218-690-9

  •  MDL number MFCD00062979

  •  PubChem Substance ID 24882791

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alcohols, Aliphatic, Alphabetical, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard, for terpene analysis
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −50±1°, c = 10% in ethanol
optical purity   enantiomeric ratio: ≥99:1 (GC)
shelf life   (limited shelf life, expiry date on the label)
mp   ~43 °C

Description

Other Notes

Important resolving agent for acids, recent applications include1; Crystallization-induced asymmetric transformation of malonates 2; Chiral auxiliary 3,4

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

(±)-Menthol

racemic, ≥98.0% (GC)

(+)-Menthol

puriss. p.a., terpene standard for GC, ≥99.0% (sum of enantiomers, GC)

(−)-Menthol

primary reference standard

(−)-Menthol

puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99%

<SC>L</SC>-Menthol
W266590L-Menthol

≥99%, FCC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?