64161 Fluka

Glycidyl methacrylate

purum, ≥97.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: 2,3-Epoxypropyl methacrylate, Methacrylic acid 2,3-epoxypropyl ester

  • CAS Number 106-91-2

  • Empirical Formula (Hill Notation) C7H10O3

  • Molecular Weight 142.15

  •  Beilstein Registry Number 2506

  •  EC Number 203-441-9

  •  MDL number MFCD00005137

  •  PubChem Substance ID 24883127

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
grade   purum
assay   ≥97.0% (GC)
contains   ~0.005% hydroquinone monomethyl ether as stabilizer
refractive index   n20/D 1.449(lit.)
  n20/D 1.450
bp   189 °C(lit.)
density   1.075 g/mL at 20 °C
  1.042 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Acrylic acid

anhydrous, contains 180-200 ppm MEHQ as inhibitor, 99%

Ethylene glycol dimethacrylate

98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor

Glycidyl methacrylate

97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor

Glycidyl methacrylate

≥97.0% (GC)

Methyl methacrylate

contains ≤30 ppm MEHQ as inhibitor, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?