-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym:
2,4-
CAS Number 50632-57-0
Empirical Formula (Hill Notation) C17H14O3
Molecular Weight 266.29
Beilstein Registry Number 1288529
MDL number MFCD00036834
PubChem Substance ID 24883558
| Related Categories | Alphabetical, Amines, Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, |
| assay | ≥98.0% (HPLC) |
| fluorescence | λex 384 nm; λem 472 nm in acetonitrile (after derivatization with hexylamine) |
| suitability | suitable for fluorescence |
| storage temp. | 2-8°C |
Reagent for the derivatization of primary and secondary amines for HPLC; highly fluorescent derivatives are formed1,2; Pre-column derivatization of amines3; Fluorescent labeling of proteins before SDS-PAGE4
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?