Synonym:
(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-β-
CAS Number 52286-58-5
Empirical Formula (Hill Notation) C42H72O14
Molecular Weight 801.01
MDL number MFCD00210509
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, G, Nutrition Research, |
| grade | analytical standard |
| assay | ≥95.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?