66490 Fluka

Methylcyclopentane

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 96-37-7

  • Linear Formula C5H9CH3

  • Molecular Weight 84.16

  •  Beilstein Registry Number 1900214

  •  EC Number 202-503-2

  •  MDL number MFCD00001382

  •  PubChem Substance ID 24884699

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
vapor pressure   232.8 mmHg ( 37.7 °C)
assay   ≥99.5% (GC)
autoignition temp.   624 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   8.35 %
refractive index   n20/D 1.409(lit.)
  n20/D 1.410
bp   72 °C(lit.)
mp   −142 °C(lit.)
density   0.748 g/mL at 20 °C
  0.749 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

2-Methylpentane

analytical standard

2-Methylpentane

≥99%

3-Methylpentane

analytical standard

Methylcyclohexane

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?