Synonym: 4,4′-Diamino-3,3′-dichlorodiphenylmethane
CAS Number 101-14-4
Linear Formula CH2[C6H3(Cl)NH2]2
Molecular Weight 267.15
Beilstein Registry Number 1882318
EC Number 202-918-9
MDL number MFCD00047829
PubChem Substance ID 24884819
| Related Categories | 2002/61/EC, Alphabetical Index of Analytical Standards, Amines, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for environmental analysis |
| shelf life | (limited shelf life, expiry date on the label) |
| bp | 202-214 °C/0.3 mmHg(lit.) |
| mp | 102-107 °C(lit.) |
Standard for determining aromatic amines released from azo-dyes in commodities1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?