Synonym:
3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside,
Myricetin 3-O-α-
CAS Number 17912-87-7
Empirical Formula (Hill Notation) C21H20O12
Molecular Weight 464.38
EC Number 241-856-7
MDL number MFCD00016930
| Related Categories | Alphabetical, Analytical Standards, Analytical/Chromatography, Arctostaphylos, Betula, |
| grade | analytical standard |
| assay | ≥95.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Bottomless glass bottle. Contents are inside inserted fused cone.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?