67870 Fluka

4-Methylmorpholine

purum, ≥98.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: NMM

  • CAS Number 109-02-4

  • Empirical Formula (Hill Notation) C5H11NO

  • Molecular Weight 101.15

  •  Beilstein Registry Number 102719

  •  EC Number 203-640-0

  •  MDL number MFCD00006175

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, C4 to C10, Chemical Synthesis, Heterocyclic Building Blocks, Morpholines/Thiomorpholines More...
vapor density   >1 (vs air)
vapor pressure   18 mmHg ( 20 °C)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.435(lit.)
  n20/D 1.435
bp   115-116 °C/750 mmHg(lit.)
mp   −66 °C(lit.)
density   0.918 g/mL at 20 °C
  0.92 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

4-Methylmorpholine

purified by redistillation, ≥99.5%

4-Methylmorpholine

ReagentPlus®, 99%

4-Methylmorpholine

BioXtra, suitable for protein sequencing, ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?