67886 Fluka

Pyrase Strips

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Pyrrolidonyl Peptidase Strips

Properties

Related Categories Discs and Strips, Identification Tests & Reagents, Microbiology More...
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
storage temp.   2-8°C

Description

Components

The kit contains plastic strips with a paper zone saturated with chromogenic substrate for the detection of pyrrolidonyl peptidase and a bottle containing a developing reagent.

Application

Diagnostic test for the rapid differentiation of Enterococci from the group D Streptococci and differentiation of Streptococcus pyogenes from other haemolytic Streptococci.
It is an enzymatic test for the detection of pyrrolidonyl peptidase (pyrase). Pyrase is an enzyme produced by all common species of Enterococci and by 98% of S. pyogenes strains. Detection of pyrase is a sensitive, reliable and rapid method of wide application. It is substantially more efficient than the bacitracine disc tests (Fluka 08382) used for the differentiation of S. pyogenes strains and the tests used for grouping of Enterococci. Strip can also be used for the identification of Gram-positive genera, catalase negative cocci, and coccobacilli.
The result of the reaction is read after 2 min.

Packaging

50 test strips, 1.0 mL reagent

Price and Availability

Customers Also Viewed

Bacitracin

from Bacillus licheniformis, ≥50,000 U/g

Hydrogen peroxide solution

3%, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?