Synonym: Anaerobe gas generation bags
Popular Documents: Datasheet (PDF)
| Related Categories | Anaerobic Equipment, Core Bioreagents, Labware, Life Science Reagents for Cloning, Microbiology Equipment, |
| grade | for microbiology |
The anaerobic atmosphere generation bags are highly innovative products designed to safely and speedily create optimum conditions for the growth of anaerobic organisms. No water is added, no hydrogen is produced, and no catalyst is required. The anaerobic atmosphere is completed within 30 minutes.
Each box contains 10 paper sachets which are individually foil packed; 1 Product Insert, the sachet is designed for use in 2.5 liter jars (Catalog No. 28029).
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?