EMAIL THIS PAGE TO A FRIEND

68390 Fluka

2-Methyl-3-pentanol

purum, ≥98.0% (GC)

Synonym: Ethyl isopropyl carbinol

Purchase

Properties

grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.417(lit.)
  n20/D 1.417
bp   128 °C(lit.)
density   0.819 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Risk Statements (Europe) 
10
Safety Statements (Europe) 
23-24/25
RIDADR 
UN 1987 3/PG 3
WGK Germany 
3
RTECS 
SA7400000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 1,IV,1716

Aldrich MSDS 1, 1285:C / Corp MSDS 1 (2), 2403:C / FT-IR 1 (1), 123:C / FT-IR 2 (1), 178:A / FT-NMR 1 (1), 185:B / IR-Spectra (2), 71:G / IR-Spectra (3), 74:A / NMR-Reference 2 (1), 115:C / RegBook 1 (1), 133:H / Structure Index 1, 16:B:1 / Vapor Phase 3, 174:C

Related Products

replaced by

Product #

Description

Add to Cart

129976 2-Methyl-3-pentanol, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?