Synonym: Oxalic acid solution
CAS Number 144-62-7
Linear Formula HOOCCOOH
Molecular Weight 90.03
EC Number 231-791-2
MDL number MFCD00002573
PubChem Substance ID 24885825
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Reagents, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules, |
| concentration | 0.1 M (COOH)2 (0.2N) |
| Gene Information | human ... SRC(6714) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?