Synonym:
1-
CAS Number 85-86-9
Empirical Formula (Hill Notation) C22H16N4O
Molecular Weight 352.39
Beilstein Registry Number 931185
Colour Index Number 26100
EC Number 201-638-4
MDL number MFCD00003905
| Related Categories | All Alphabetically Sorted, All alphabetical sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for food analysis |
| assay | ≥96.0% (HPLC) |
| mp | 199 °C (dec.)(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?