CAS Number 89-25-8
Empirical Formula (Hill Notation) C10H10N2O
Molecular Weight 174.20
Beilstein Registry Number 609575
EC Number 201-891-0
MDL number MFCD00003138
PubChem Substance ID 24885706
Popular Documents: FTNMR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Coupling, Derivatization Reagents, |
| grade | purum |
| assay | ≥98.0% (NT) |
| bp | 287 °C/265 mmHg(lit.) |
| mp | 126-128 °C(lit.) |
| 126-128 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?