Synonym:
4-
CAS Number 633-96-5
Empirical Formula (Hill Notation) C16H11N2NaO4S
Molecular Weight 350.32
Beilstein Registry Number 3898201
Colour Index Number 15510
EC Number 211-199-0
MDL number MFCD00011657
| Related Categories | All Alphabetically Sorted, All alphabetical sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| assay | ≥98.0% (HPLC) |
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?