Synonym: 4-Methoxyfuro[3,2-g]benzopyrane-7-one, 5-Methoxypsoralen, Bergapten
CAS Number 484-20-8
Empirical Formula (Hill Notation) C12H8O4
Molecular Weight 216.19
Beilstein Registry Number 19560
EC Number 207-604-5
MDL number MFCD00010272
| Related Categories | A - D, A-F, All Alphabetically Sorted, Alphabetical, Analytical Standards, |
| grade | analytical standard |
| assay | ≥99.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | 189-193 °C |
| 190-193 °C(lit.) | |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?