CAS Number 50-07-7
Empirical Formula (Hill Notation) C15H18N4O5
Molecular Weight 334.33
Beilstein Registry Number 7231816
MDL number MFCD00078109
| Related Categories | Antibiotics, Antibiotics A to Z, Antibiotics G-M, Antibiotics by Application, Antineoplastic and Immunosuppressive Antibiotics, |
| storage temp. | 2-8°C |
Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial to gram positive, gram negative, acid-fast bacilli.
Vial contains 2 mg mitomycin C and 48 mg NaCl (increases solubility).
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?