Synonym: Molecular Sieve UOP Type 3Å
CAS Number 308080-99-1
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
MDL number MFCD00147627
| Related Categories | Adsorbents, Filter Aids and Drying Agents, Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Drying Agents, |
| form | rod |
| size | 1/16 in. |
| loss | ≤3.0% loss on ignition, 900°C |
| pore size | 3 Å pore diameter |
Customers Also Viewed
beads
powder
beads, 4-8 mesh
beads, 8-12 mesh
pellets, 3.2 mm
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?