69899 Fluka

Monochlorobimane

suitable for fluorescence, ≥95% (HPCE)

DOWNLOAD MSDS (PDF)

Synonym: Chlorobimane

  • CAS Number 76421-73-3

  • Empirical Formula (Hill Notation) C10H11ClN2O2

  • Molecular Weight 226.66

  •  Beilstein Registry Number 4440901

  •  MDL number MFCD00077379

  •  PubChem Substance ID 24886000

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Indicators and Probes, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains,
assay   ≥95% (HPCE)
mp   135-136 °C(lit.)
solubility   DMF: soluble
  DMSO: soluble
  acetonitrile: soluble
  methanol: soluble
fluorescence   λex 380 nm; λem 461 nm in methanol
  λex 390 nm; λem 478 nm in 0.1 M phosphate pH 7.5 (after derivatization with glutathione)
suitability   suitable for fluorescence

Description

Application

Cell-permeant probe for quantifying glutathione levels in cells.1 Suitable for the detection of thiol-groups.

General description

Contains up to 15% of an isomer (NMR)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Bromobimane

≥97%

Bromobimane

BioReagent, suitable for fluorescence, ≥95% (HPCE)

sufficient for 200 multiwell tests

lyophilized powder, ≥25 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?