70012 Fluka
AQUANAL-professional SPECTRO 1000
single-beam-spectrometer
DOWNLOAD MSDS (PDF)
Description
Application
suitable for all AQUANAL-professional tests
Other Notes
16 and 24 mm cyclic tubes and 10 to 50 mm rectangular tubes, region of wavelength 330 - 900 nm
absorption/ transmission, taking of spectra, several given methods
Legal Information
AQUANAL is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Customers Also Viewed
for evaluation of the COD cuvet tests see 37735, 37736, 37737, 37738
AQUANAL<SUP>™</SUP>-plus Spectro Hazen
compact photometer for the determination Hazen (Pt-Co-Scale; APHA)
Genesys 20 spectrophotometer
With U.S. cord
Genesys 20 spectrophotometer
With Euro cord
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?