Synonym: MPO, Peroxidase, myelo
CAS Number 9003-99-0
MDL number MFCD00131641
| Related Categories | 1.11.x.x Peroxide as receptor, 1.x.x.x Oxidoreductases, Application Index, Biochemicals and Reagents, Diagnostic and Analytical Enzymes, |
| form | powder |
| quality | lyophilized |
| mol wt | Mr 135000-150000 |
| color | white |
| shipped in | dry ice |
| storage temp. | −20°C |
lyophilized from 20 mM sodium acetate buffer, pH 6,0
Review1
1 U corresponds to the amount of enzyme which will produce an increase in absorption (A470) of 1.0 per minute at pH 7.0 and 25°C, calculated from the initial rate of reaction using guaiacol (Fluka No. 50880) as substrate; total reaction volume: 3.
Customers Also Viewed
lyophilized powder, ≥50 units/mg protein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?