70129 Fluka

Methyl myristate

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Methyl tetradecanoate, Myristic acid methyl ester

  • CAS Number 124-10-7

  • Linear Formula CH3(CH2)12COOCH3

  • Molecular Weight 242.40

  •  Beilstein Registry Number 1773739

  •  EC Number 204-680-1

  •  MDL number MFCD00008983

  •  PubChem Substance ID 24886058

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Chromatography,
grade   analytical standard
assay   ≥99.5% (GC)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.436(lit.)
  n20/D 1.438
bp   323 °C(lit.)
mp   18 °C(lit.)
density   0.855 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Methyl dodecanoate

analytical standard

Methyl myristate

≥99% (GC)

Methyl myristate

≥98%

Methyl oleate

analytical standard

Methyl palmitate

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?