EMAIL THIS PAGE TO A FRIEND

70139 Fluka

Potato Dextrose Agar

for microbiology

Synonym: Potato Glucose Agar

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Aspergillus, Candida,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 15 g/L
  dextrose, 20 g/L
  potato extract, 4 g/L
final pH   5.6±0.2 (25 °C)
suitability   nonselective for Aspergillus
  nonselective for Candida
  nonselective for Molds and Yeasts (General Media)
  nonselective for Penicilliums
  nonselective for Pichia
  nonselective for Saccharomyces
  nonselective for Zygosaccharomyces

Description

Application

Recommended for the isolation and enumeration of yeasts and molds in dairy products and food.

Other Notes

Studies on contamination of yogurt by yeast1,2; For maintaining the stock culture of Rhodotorula glutinis for the production of extracellular lipase3; Identification of Aspergillus niger from infected onion tissue4

Price and Availability

Additional Products to Explore

for molecular biology

Glycerol

for molecular biology, ≥99%

Sodium chloride

BioXtra, ≥99.5% (AT)

Sodium hydroxide

ACS reagent, ≥97.0%, pellets

Sucrose
S7903Sucrose

BioXtra, ≥99.5% (GC)

Customers Also Viewed

for microbiology

for microbiology

for microbiology, ready to use media in the bottle

Potato Dextrose Agar

for microbiology

Potato Dextrose Broth

for microbiology

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Yeasts

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.
Jvo Siegrist
Analytix 2009, 5, 7.
Keywords: Antibiotics, Enzyme-linked immunosorbent assay, Fermentation, Microbiology, Molecular biology, Polymerase chain reaction, Reproduction, Vitamins

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. M. Dublin Green, S.N. Ibe J. Food Prot. 50, 193, (1987)

2. R.E. Beever, E.G. Bollard J. Gen. Microbiol. 60, 273, (1970)

3. D. Papaparaskevas et al. Biotechnol. Lett. 14, 397, (1992)

4. A. Behere et al. Enzyme Microb. Technol. 15, 158, (1993)

Conservation and diversity of seed associated endophytes in Zea across boundaries of evolution, ethnography and ecology. Johnston-Monje D PLoS ONE 6(6), e20396, (2011)

Loading...


Calcineurin-responsive zinc finger transcription factor CRZ1 of Botrytis cinerea is required for growth, development, and full virulence on bean plants. Schumacher J Eukaryotic Cell 7(4), 584-601, (2008)

Loading...


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?