• USA Home
  • 70157 - Thioglycollate Broth (USP Alternative)

EMAIL THIS PAGE TO A FRIEND

70157 Fluka

Thioglycollate Broth (USP Alternative)

for microbiology

Synonym: Alternative Thioglycollate Medium, Fluid Thioglycollate Medium, NIH Thioglycollate Broth, TG Broth

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Anaerobic Bacteria, Aspergillus,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   L-cystine, 0.5 g/L
  dextrose, 5.5 g/L
  pancreatic digest of casein, 15.0 g/L
  sodium chloride, 2.5 g/L
  sodium thioglycolate, 0.5 g/L
  yeast extract, 5.0 g/L
final pH   7.1±0.2 (25 °C)
suitability   Anaerobic Bacteria
  nonselective for Aspergillus
  nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Bacteroides
  nonselective for Candida
  nonselective for Clostridia
  nonselective for Escherichia coli and Coliforms
  nonselective for Pseudomonas
  nonselective for Staphylococcus

Description

Application

A medium for sterility tests and the cultivation of microaerophilic and anaerobic organisms.

Other Notes

Sterility tests1,2

Price and Availability

Customers Also Viewed

for microbiology

Sodium thioglycolate

≥96.5% (iodometric)

Thioglycollate medium

Microbiologically tested

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Clostridia Diagnostic

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism. Most Clostridia cannot grow under aerobic conditions and even can be killed by exposure...
Jvo Siegrist
AnalytiX Volume 7 Article 2
Keywords: Aerobic, Anaerobic, Clinical, Degradations, Environmental, Fermentation, Germination, Gram stain, Indicators, Metabolism, Microbiology, Pharmaceutical, Vitamins

Pseudomonas Media and Tests

Pseudomonas are motile (one or more polar flagella), rod shaped and aerobe Gram-negative bacteria. They are found almost everywhere, in soil, water, plants and animals. In most cases it is not pathog...
Jvo Siegrist, Product Manager Microbiology, ivo.siegrist@sial.com
Analytix Volume 2007 Article 5
Keywords: Anaerobic, Biochemistry, Cosmetics, Diagnostic, Fermentation, Glycolysis, Metabolism, Microbiology, Pentose phosphate pathway, Pharmaceutical, Respiratory

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. N.I.H. Memorandum, (1955)

2. United States Pharmacopeia 18th ed.,, 851, (1970)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?