70289 Fluka

Sodium 2-naphthalenesulfonate

for ion pair chromatography, ≥99.0% (HPLC)

DOWNLOAD MSDS (PDF)

Synonym: 2-Naphthalenesulfonic acid sodium salt

  • CAS Number 532-02-5

  • Empirical Formula (Hill Notation) C10H7NaO3S

  • Molecular Weight 230.22

  •  Beilstein Registry Number 3659024

  •  EC Number 208-523-8

  •  MDL number MFCD00064186

  •  PubChem Substance ID 24886005

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Basic Ion Pair Reagents (Cationic), Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (HPLC)
λ   10 % in H2O (hot)
UV absorption   λ: 380 nm Amax: 0.1
  λ: 390 nm Amax: 0.05
  λ: 400 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for filter test

Price and Availability

Customers Also Viewed

2-<WBR>Naphthalenesulfonic acid

technical grade, 70%

Sodium 1-<WBR>naphthalenesulfonate

technical grade, 80%

Sodium 2-<WBR>naphthalenesulfonate

technical grade, 90%

Sodium 2-<WBR>naphthalenesulfonate

technical, ≥90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?