70290 Fluka

Sodium 2-naphthalenesulfonate

technical, ≥90%

DOWNLOAD MSDS (PDF)

Synonym: 2-Naphthalenesulfonic acid sodium salt

  • CAS Number 532-02-5

  • Empirical Formula (Hill Notation) C10H7NaO3S

  • Molecular Weight 230.22

  •  Beilstein Registry Number 3659024

  •  EC Number 208-523-8

  •  MDL number MFCD00064186

  •  PubChem Substance ID 24886006

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
grade   technical
assay   ≥90%

Description

Other Notes

Used for ion-pair chromatography of amines and amino acid derivatives with UV-detection1

Price and Availability

Customers Also Viewed

Sodium 2-<WBR>naphthalenesulfonate

for ion pair chromatography, ≥99.0% (HPLC)

Sodium 2-<WBR>naphthalenesulfonate

technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?