EMAIL THIS PAGE TO A FRIEND

70480 Fluka

α-Naphtholbenzein

indicator (pH 8.2-10.0)

Synonym: 4,4′-(α-Hydroxybenzylidene)di-1-naphthol, p-naphtholbenzein

  • CAS Number 145-50-6

  • Empirical Formula (Hill Notation) C27H18O2

  • Molecular Weight 374.43

  •  Beilstein Registry Number 2628063

  •  EC Number 205-656-3

  •  PubChem Substance ID 24886093

  • Popular Documents:  FTIR (PDF)  

Purchase

Price and Availability

TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells


Customers Also Viewed

α-Naphtholbenzein

indicator grade

Crystal violet solution

0.5% in anhydrous acetic acid, indicator for non-aqueous titration

Potassium hydroxide solution

volumetric, 0.1 M KOH in isopropanol (0.1N), according to DIN 51558-1

Quinaldine Red

Dye content 95 %

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 6,1150

Corp MSDS 1 (2), 2504:D / FT-IR 2 (2), 1922:B / RegBook 1 (1), 1319:L / Structure Index 1, 202:D:2

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?