70721 Fluka

N-(1-Naphthyl)ethylenediamine dihydrochloride

purum, ACS reagent, ≥97.0% (calc. based on dry substance, AT)

DOWNLOAD MSDS (PDF)

Synonym: 2-(1-Naphthylamino)ethylamine dihydrochloride

  • CAS Number 1465-25-4

  • Linear Formula C10H7NHCH2CH2NH2 · 2HCl

  • Molecular Weight 259.17

  •  Beilstein Registry Number 3707471

  •  EC Number 215-981-2

  •  MDL number MFCD00012556

  • Popular Documents:  FTIR (PDF)  

Properties

grade   ACS reagent
  purum
assay   ≥97.0% (calc. based on dry substance, AT)
ign. residue   ≤0.1%
mp   194-198 °C (dec.)(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

<I>N</I>-<WBR>(1-<WBR>Naphthyl)<WBR>ethylenediamine dihydrochloride

for determination of sulfonamide and nitrite, ACS reagent, ≥98%

<I>N</I>-<WBR>(1-<WBR>Naphthyl)<WBR>ethylenediamine dihydrochloride monomethanolate

for spectrophotometric det. of nitrate and nitrite, ≥99.0%

Sulfanilamide

≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?