Synonym:
1-
CAS Number 84-86-6
Linear Formula H2NC10H6SO3H
Molecular Weight 223.25
Beilstein Registry Number 1971299
EC Number 201-567-9
MDL number MFCD00004027
PubChem Substance ID 24886126
Popular Documents: FTIR (PDF)
| Related Categories | Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More... |
| grade | purum |
| assay | ≥97.0% (T) |
| mp | ≥300 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?