71430 Fluka

Sodium cyanide

purum p.a., ≥96.0% (AT)

DOWNLOAD MSDS (PDF)
  • CAS Number 143-33-9

  • Linear Formula NaCN

  • Molecular Weight 49.01

  •  Beilstein Registry Number 3587243

  •  EC Number 205-599-4

  •  MDL number MFCD00003523

Properties

vapor density   1.7 (vs air)
vapor pressure   1 mmHg ( 817 °C)
grade   purum p.a.
assay   ≥96.0% (AT)
mp   563.7 °C(lit.)
anion traces   chloride (Cl-): ≤2000 mg/kg
  sulfate (SO42-): ≤500 mg/kg
cation traces   Ca: ≤100 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤1000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Customers Also Viewed

Sodium cyanide

puriss. p.a., ACS reagent, ≥97.0% (AT)

Sodium cyanide

ACS reagent, ≥95.0%

Sodium cyanide

reagent grade, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?