71435 Fluka

Sodium cyanoborohydride

purum, ≥95.0% (RT)

DOWNLOAD MSDS (PDF)

Synonym: Sodium cyanotrihydridoborate

  • CAS Number 25895-60-7

  • Linear Formula NaBH3CN

  • Molecular Weight 62.84

  •  Beilstein Registry Number 4152656

  •  MDL number MFCD00003516

Properties

Related Categories Borohydrides, Chemical Synthesis, Reduction, Synthetic Reagents
grade   purum
assay   ≥95.0% (RT)
mp   >242 °C (dec.)(lit.)

Description

Other Notes

Review1,2; selctive reductions with NaCNBH3, modified with zinc chloride3; with zinc iodide4

Price and Availability

Customers Also Viewed

Formaldehyde-d<SUB>2</SUB> solution

~20 wt. % in D2O, 98 atom % D

Sodium cyanoborodeuteride

96 atom % D, 98% (CP)

Sodium cyanoborohydride

reagent grade, 95%

Sodium cyanoborohydride solution

5.0 M in 1 M NaOH

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?