71443 Fluka

Sodium decyl sulfate

for ion pair chromatography, ≥99.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: Decyl sulfate sodium salt

  • CAS Number 142-87-0

  • Linear Formula CH3(CH2)9OSO3Na

  • Molecular Weight 260.33

  •  Beilstein Registry Number 3576860

  •  EC Number 205-568-5

  •  MDL number MFCD00041881

  •  PubChem Substance ID 24886194

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Basic Ion Pair Reagents (Cationic), Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (T)
λ   10 % in H2O
UV absorption   λ: 210 nm Amax: 0.05
  λ: 220 nm Amax: 0.04
  λ: 230 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for filter test

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Sodium decyl sulfate

puriss. p.a., for surfactant analysis, ≥99.0% (T)

Sodium octyl sulfate

for ion pair chromatography, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?