• USA Home
  • 71463 - Sodium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
71463 Fluka

Sodium hydroxide solution

volumetric, 1 M NaOH (1N)

Purchase

Properties

Related Categories Acids & Bases, Analytical Reagents, Analytical/Chromatography, Base Solutions, Bases,
vapor pressure   3 mmHg ( 37 °C)
quality   volumetric
shelf life   limited shelf life
calibrated against   potassium hydrogen phthalate
concentration   1 M NaOH (1N)

Description

General description

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1824 8 / PGIII
WGK Germany 
1
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?