71607 Fluka

Ethyl isovalerate

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 108-64-5

  • Linear Formula (CH3)2CHCH2COOC2H5

  • Molecular Weight 130.18

  •  Beilstein Registry Number 1744677

  •  EC Number 203-602-3

  •  MDL number MFCD00009203

  •  PubChem Substance ID 24886231

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories A-F, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alpha Sort, Alphabetical Index of Analytical Standards,
grade   analytical standard
vapor pressure   7.5 mmHg ( 20 °C)
assay   ≥99.7% (GC)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.396(lit.)
  n20/D 1.397
bp   131-133 °C(lit.)
mp   −99 °C(lit.)
density   0.864 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Ethyl 2-methylbutyrate

≥98%, FCC, FG

Ethyl isovalerate

≥98%, FCC, FG

Ethyl isovalerate

natural, ≥98%, FCC, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?