• USA Home
  • 71726 - Sodium dodecyl sulfate

EMAIL THIS PAGE TO A FRIEND

71726 Fluka

Sodium dodecyl sulfate

for ion pair chromatography, ≥99.0%

Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Purchase

Properties

Related Categories Alkyl Sulfates, Analytical Reagents, Analytical/Chromatography, Anionic Detergents, Antigen-Vaccine Preparation,
grade   for ion pair chromatography
description   anionic
assay   ≥99.0% (GC)
  ≥99.0%
mp   204-207 °C(lit.)
λ   10 % in H2O
UV absorption   λ: 210 nm Amax: 0.1
  λ: 220 nm Amax: 0.06
  λ: 230 nm Amax: 0.04
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Description

Application

Anionic detergent

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells


Customers Also Viewed

Sodium dodecyl sulfate

dust-free pellets, ≥99.0% (GC)

Sodium dodecyl sulfate

BioXtra, ≥99.0% (GC)

Sodium dodecyl sulfate

ACS reagent, ≥99.0%

Sodium dodecyl sulfate

ReagentPlus®, ≥98.5% (GC)

Sodium dodecyl sulfate

BioUltra, for molecular biology, ≥99.0% (GC)

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37-39
RIDADR 
UN 2926 4.1/PG 2
WGK Germany 
2
RTECS 
WT1050000
Flash Point(F) 
338 °F
Flash Point(C) 
170 °C

Protocols & Articles

Articles

Determination of Water Content in Texpon 100 Using Karl Fischer Titration

Cosmetic products are very often preparations with a very high water content which must be exactly determined. This requires precise sampling, weighing and administration of the substance. The sample...
Keywords: Sample preparations, Titrations

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Measurement of molecular weight by gel electrophoresis. T.B. Nelsen, J.A. Reynolds Meth. Enzymol. 48, 3, (1978)

Loading...


Analysis of drugs in the presence of serum albumin by LC with eluants containing surfactants. R.A. Grohs Anal. Chem. 63, 384, (1991)

Loading...


A rapid electrophoretic procedure for the detection of SDS-released oncorna-viral RNA using polyacrylamide-agarose gels. Harewood, K. and Wolff, J.S. Anal. Biochem. 55, 573, (1973)

Loading...


Cleavage of structural proteins during the assembly of the head of bacteriophage T4. Laemmli, U.K. Nature 227, 680-685, (1970)

Loading...


Biofilm development and enhanced stress resistance of a model, mixed-species community biofilm. Lee KW, Periasamy S, Mukherjee M, et al. ISME J. 8(4), 894-907, (2014)

Loading...


Loss of mural cells leads to wall degeneration, aneurysm growth, and eventual rupture in a rat aneurysm model. Marbacher S, Marjamaa J, Bradacova K, et al. Stroke 45(1), 248-54, (2014)

Loading...


Exploring the molecular basis of insecticide resistance in the dengue vector Aedes aegypti: a case study in Martinique Island (French West Indies). Sébastien Marcombe et al BMC Genomics 10, 494, (2009)

Loading...


Straightforward palladium-mediated synthesis and biological evaluation of benzo[j]phenanthridine-7,12-diones as anti-tuberculosis agents. Davie Cappoen et al Eur. J. Med. Chem. 48, 57-68, (2012)

Loading...


Characterization of UDP-glucose dehydrogenase and UDP-glucose pyrophosphorylase mutants of Proteus mirabilis: defectiveness in polymyxin B resistance, swarming, and virulence. Sin-Sien Jiang et al Antimicrob. Agents Chemother. 54, 2000-9, (2010)

Loading...


Fate and effects of the surfactant sodium dodecyl sulfate. Singer MM and Tjeerdema RS Rev. Environ. Contam. Toxicol. 133, 95-149, (1993)

Loading...


[Complexes formed between sodium dodecyl sulfate and protein polypeptide with special reference to SDS-polyacrylamide gel electrophoresis (author's transl)]. Takagi T, Shirahama K, and Kubo K Tanpakushitsu Kakusan Koso. 21(10), 811-29, (1976)

Loading...


[Sodium dodecyl sulfate (SDS)--for the users in the field of biochemistry (author's transl)]. Tsujii K and Takagi T Tanpakushitsu Kakusan Koso. 19(9), 705-16, (1974)

Loading...


Effect of sodium dodecyl sulfate (SDS) on stress response in the Mediterranean mussel (Mytilus Galloprovincialis): regulatory volume decrease (Rvd) and modulation of biochemical markers related to oxidative stress. Messina CM, Faggio C, Laudicella VA, et al. Aquat. Toxicol. 157, 94-100, (2014)

Loading...


Influence of mounthrinse solutions on the color stability and microhardness of a composite resin. Lepri CP, Ribeiro MV, Dibb A, et al. Int. J. Esthet. Dent. 9(2), 238-46, (2014)

Loading...


Use of statistical modeling to predict the effect of formulation composition on conditioning shampoo performance. Lepilleur C, Giovannitti-Jensen A, and Kyer C J. Cosmet. Sci. 64(6), 411-27, (2013)

Loading...


[Antimicrobial activity of stable silver nanoparticles of a certain size]. Mukha IuP, Eremenko AM, Smirnova NP, et al. Prikl. Biokhim. Mikrobiol. 49(2), 215-23, (2013)

Loading...


Refolding of SDS-denatured proteins using amphipathic cosolvents and osmolytes. Roussel G, Tinti E, Perpète E, et al. Curr. Protoc. Protein Sci. Chapter 28, Unit28.5, (2013)

Loading...


"Mixed" anionic and non-ionic micellar liquid chromatography for high-speed radiometabolite analysis of positron emission tomography radioligands. Nakao R and Halldin C J. Chromatogr. A 1281, 54-9, (2013)

Loading...


Effect of curing light distance and different mouthwashes on the sorption and solubility of a nanofilled composite. de Azevedo Miranda D, dos Santos Bertoldo CE, Ambrosano GM, et al. Eur. J. Esthet. Dent. 8(1), 88-102, (2013)

Loading...


Capillary electrophoresis in presence of sodium dodecyl sulfate and a sieving medium. Takagi T Electrophoresis 18(12-13), 2239-42, (1997)

Loading...


New plasmid-mediated fluoroquinolone efflux pump, QepA, found in an Escherichia coli clinical isolate. Kunikazu Yamane et al Antimicrob. Agents Chemother. 51, 3354-60, (2007)

Loading...


Micelle-induced multiple performance improvement of fluorescent probes for H2S detection. Tian H, Qian J, Bai H, et al. Anal. Chim. Acta 768, 136-42, (2013)

Loading...


Disclosure of a structural milieu for the proximity ligation reveals the elusive nature of an active chromatin hub. Gavrilov AA, Gushchanskaya ES, Strelkova O, et al. Nucleic Acids Res. 41(6), 3563-75, (2013)

Loading...


Novel cationic lipophilic peptides for oligodeoxynucleotide delivery. Enoch Chan et al Bioorg. Med. Chem. 15, 4091-7, (2007)

Loading...


Comparative static curing versus dynamic curing on tablet coating structures. Gendre C, Genty M, Fayard B, et al. Int. J. Pharm. 453(2), 448-53, (2013)

Loading...


Incorporation of polyamidoamine sweeping and electrokinetic supercharging for in-line DNA fragment preconcentration. Tian J, Qiao J, Gao J, et al. J. Chromatogr. A 1280, 112-6, (2013)

Loading...


Loop sequence dictates the secondary structure of a human membrane protein hairpin. Nadeau VG and Deber CM Biochemistry 52(14), 2419-26, (2013)

Loading...


Role of AbeS, a novel efflux pump of the SMR family of transporters, in resistance to antimicrobial agents in Acinetobacter baumannii. Vijaya Bharathi Srinivasan et al Antimicrob. Agents Chemother. 53, 5312-6, (2009)

Loading...


Prediction of micelle/water and liposome/water partition coefficients based on molecular dynamics simulations, COSMO-RS, and COSMOmic. Ingram T, Storm S, Kloss L, et al. Langmuir 29(11), 3527-37, (2013)

Loading...


Reduced biocide susceptibility in Candida albicans biofilms. Jeniel E Nett et al Antimicrob. Agents Chemother. 52, 3411-3, (2008)

Loading...


LmrS is a multidrug efflux pump of the major facilitator superfamily from Staphylococcus aureus. Jody L Floyd et al Antimicrob. Agents Chemother. 54, 5406-12, (2010)

Loading...


Comparison of complexes formed by a crustacean and a vertebrate trypsin with bovine pancreatic trypsin inhibitor - the key to achieving extreme stability? Molnár T, Vörös J, Szeder B, et al. FEBS J. 280(22), 5750-63, (2013)

Loading...


Cloning, nucleotide sequencing, and analysis of the AcrAB-TolC efflux pump of Enterobacter cloacae and determination of its involvement in antibiotic resistance in a clinical isolate. Astrid Pérez et al Antimicrob. Agents Chemother. 51, 3247-53, (2007)

Loading...


Anti-inflammatory activity of verminoside from Kigelia africana and evaluation of cutaneous irritation in cell cultures and reconstituted human epidermis. Patrizia Picerno et al J. Nat. Prod. 68, 1610-4, (2005)

Loading...


Substrate competition studies using whole-cell accumulation assays with the major tripartite multidrug efflux pumps of Escherichia coli. Christopher A Elkins et al Antimicrob. Agents Chemother. 51, 923-9, (2007)

Loading...


Mechanism of electrolyte-induced brightening in single-wall carbon nanotubes. Duque JG, Oudjedi L, Crochet JJ, et al. J. Am. Chem. Soc. 135(9), 3379-82, (2013)

Loading...


The role of amphiphilicity and negative charge in glycoprotein 41 interactions in the hydrophobic pocket. Miriam Gochin et al J. Med. Chem. 52, 4338-44, (2009)

Loading...


Fluorescent labeling of proteins with nile red and 2-methoxy-2,4-diphenyl-3(2H)-furanone: physicochemical basis and application to the rapid staining of sodium dodecyl sulfate polyacrylamide gels and Western blots. Daban JR Electrophoresis 22(5), 874-80, (2001)

Loading...


Decellularized feeders: an optimized method for culturing pluripotent cells. Lim ML, Jungebluth P, Sjöqvist S, et al. Stem Cells Transl. Med. 2(12), 975-82, (2013)

Loading...


Ion pair hollow fiber liquid-liquid-liquid microextraction combined with capillary electrophoresis-ultraviolet detection for the determination of thyroid hormones in human serum. Li P, Hu B, He M, et al. J. Chromatogr. A 1356, 23-31, (2014)

Loading...


Antigen retrieval in cells and tissues: enhancement with sodium dodecyl sulfate. Robinson JM and Vandré DD Histochem. Cell Biol. 116(2), 119-30, (2001)

Loading...


Non-targeted identification of prions and amyloid-forming proteins from yeast and mammalian cells. Kryndushkin D, Pripuzova N, Burnett BG, et al. J. Biol. Chem. 288(38), 27100-11, (2013)

Loading...


Eosinophil as a cellular target of the ocular anti-allergic action of mapracorat, a novel selective glucocorticoid receptor agonist. Monica Baiula et al Mol. Vis. 17, 3208-23, (2011)

Loading...


Interaction between zwitterionic surface activity ionic liquid and anionic surfactant: Na(+)-driven wormlike micelles. Wang X, Wang R, Zheng Y, et al. J. Phys. Chem. B 117(6), 1886-95, (2013)

Loading...


Physicochemical and antibacterial properties of surfactant mixtures with quaternized chitosan microgels. Richardson KE, Xue Z, Huang Y, et al. Carbohydr. Polym. 93(2), 709-17, (2013)

Loading...


Evaluation of the potential for diacetylenes as reporter molecules in 3D micelle gel dosimetry. Nasr AT, Olding T, Schreiner LJ, et al. Phys. Med. Biol. 58(4), 787-805, (2013)

Loading...


The detection of enzyme activity following sodium dodecyl sulfate-polyacrylamide gel electrophoresis. Bischoff KM, Shi L, and Kennelly PJ Anal. Biochem. 260(1), 1-17, (1998)

Loading...


Does CA125 binding to mesothelin impact the detection of malignant mesothelioma? Creaney J, Dick IM, Dare H, et al. Lung Cancer 80(1), 39-44, (2013)

Loading...


Influence of membrane structure on the preparation of colloidal lipid dispersions by premix membrane emulsification. Joseph S and Bunjes H Int. J. Pharm. 446(1-2), 59-62, (2013)

Loading...


Structural determinants of Tau aggregation inhibitor potency. Schafer KN, Cisek K, Huseby CJ, et al. J. Biol. Chem. 288(45), 32599-611, (2013)

Loading...


Docetaxel-loaded polylactic acid-co-glycolic acid nanoparticles: formulation, physicochemical characterization and cytotoxicity studies. Pradhan R, Poudel BK, Ramasamy T, et al. J. Nanosci. Nanotechnol. 13(8), 5948-56, (2013)

Loading...


Interface-induced disassembly of a self-assembled two-component nanoparticle system. Gao Y, Duc le T, Ali A, et al. Langmuir 29(11), 3654-61, (2013)

Loading...


Bicephalic amphiphile architecture affects antibacterial activity. Jade E LaDow et al Eur. J. Med. Chem. 46, 4219-26, (2011)

Loading...


Vesicles and reverse vesicles of an ionic liquid in ionic liquids. Rao KS, So S, and Kumar A Chem. Commun. (Camb.) 49(73), 8111-3, (2013)

Loading...


Molecular dynamics of the interaction of anionic surfactants with liposomes. Chen C, Jiang C, and Tripp CP Colloids Surf. B Biointerfaces 105, 173-9, (2013)

Loading...


In vitro reconstitution of complexes between pro-matrix metalloproteinase-9 and the proteoglycans serglycin and versican. Malla N, Berg E, Theocharis AD, et al. FEBS J. 280(12), 2870-87, (2013)

Loading...


Gamma interferon and interleukin-1 receptor 1 regulate neutrophil recruitment to the corneal stroma in a murine model of Onchocerca volvulus keratitis. Gentil K Infect. Immun. 77(4), 1606-12, (2009)

Loading...


Molecular weight standards for calibration of gel filtration and sodium dodecyl sulfate-polyacrylamide gel electrophoresis: ferritin and apoferritin. de Haen, C. Anal. Biochem. 16, 235, (1987)

Loading...


Exposure of RML scrapie agent to a sodium percarbonate-based product and sodium dodecyl sulfate renders PrPSc protease sensitive but does not eliminate infectivity. Smith JD, Nicholson EM, Foster GH, et al. BMC Vet. Res. 9, 8, (2013)

Loading...


Shotgun proteomic analysis of two Bartonella quintana strains. Fabietti A, Gaspari M, Krishnan S, et al. Proteomics 13(8), 1375-8, (2013)

Loading...


Sequence homology analysis of proteins by chemical cleavages: using a mono and two dimensional sodium dodecyl sulfate-polyacrylamide gel electrophoresis. Han KK, Richard C, Zhang GY, et al. Int. J. Biochem. 18(12), 1073-82, (1986)

Loading...


Effects of buffered vinegar and sodium dodecyl sulfate plus levulinic acid on Salmonella Typhimurium survival, shelf-life, and sensory characteristics of ground beef patties. Stelzleni AM, Ponrajan A, and Harrison MA Meat Science 95(1), 1-7, (2013)

Loading...


Different stratum corneum lipid liposomes as models to evaluate the effect of the sodium dodecyl sulfate. López O, Cócera M, de la Maza A, et al. Biochim. Biophys. Acta 1508(1-2), 196-209, (2000)

Loading...


Charged surfactants induce a non-fibrillar aggregation pathway of amyloid-beta peptide. Loureiro JA, Rocha S, and Pereira Mdo C J. Pept. Sci. 19(9), 581-7, (2013)

Loading...


Performance of different C18 columns in reversed-phase liquid chromatography with hydro-organic and micellar-organic mobile phases. Ruiz-Angel MJ, Pous-Torres S, Carda-Broch S, et al. J. Chromatogr. A 1344, 76-82, (2014)

Loading...


Effect of low concentration sodium dodecyl sulfate on the electromigration of palonosetron hydrochloride stereoisomers in micellar electrokinetic chromatography. Hu SQ, Wang GX, Guo WB, et al. J. Chromatogr. A 1342, 86-91, (2014)

Loading...


Epitope scanning indicates structural differences in brain-derived monomeric and aggregated mutant prion proteins related to genetic prion diseases. Tapella L, Stravalaci M, Bastone A, et al. Biochem. J. 454(3), 417-25, (2013)

Loading...


Formulation and in vitro and in vivo evaluation of film-coated montelukast sodium tablets using Opadry® yellow 20A82938 on an industrial scale. Zaid AN, Natour S, Qaddomi A, et al. Drug Des. Devel. Ther. 7, 83-91, (2013)

Loading...


Micellar-enhanced ultrafiltration of copper ions using sodium dodecyl sulfate and its mixture with Brij 35, Tween 80 and Triton X-100. Zhao B, Li R, Zhong J, et al. Water Sci. Technol. 67(10), 2154-9, (2013)

Loading...


Role of sol with iron oxyhydroxide/sodium dodecyl sulfate composites on Fenton oxidation of sorbed phenanthrene in sand. Park JY and Kim JH J. Environ. Manage. 126, 72-8, (2013)

Loading...


Use of surface plasmon resonance to study the adsorption of detergents on poly(dimethylsiloxane) surfaces. Gaspar A, Kecskemeti A, and Gomez FA Electrophoresis 34(8), 1249-52, (2013)

Loading...


mosR, a novel transcriptional regulator of hypoxia and virulence in Mycobacterium tuberculosis. Bassam Abomoelak et al J. Bacteriol. 191, 5941-52, (2009)

Loading...


Preparation and evaluation of Zr-beta-FeOOH for efficient arsenic removal. Sun X, Hu C, and Qu J J. Environ. Sci. (China) 25(4), 815-22, (2013)

Loading...


Effect of surfactant addition on removal of microbubbles using ultrasound. Kobayashi D, Hayashida Y, Sano K, et al. Ultrasonics 54(6), 1425-9, (2014)

Loading...


Novel Application of Cyclolipopeptide Amphisin: Feasibility Study as Additive to Remediate Polycyclic Aromatic Hydrocarbon (PAH) Contaminated Sediments. Anne Groboillot et al Int. J. Mol. Sci. 12, 1787-806, (2011)

Loading...


Structural elucidation of transmembrane transporter protein bilitranslocase: conformational analysis of the second transmembrane region TM2 by molecular dynamics and NMR spectroscopy. Roy Choudhury A, Perdih A, Zuperl S, et al. Biochim. Biophys. Acta 1828(11), 2609-19, (2013)

Loading...


Efficacy of sanitizers in reducing Salmonella on pecan nutmeats during cracking and shelling. Beuchat LR, Mann DA, and Alali WQ J. Food Prot. 76(5), 770-8, (2013)

Loading...


Concentration-dependent antagonistic persuasion of SDS and naphthalene derivatives on the fibrillation of stem bromelain. Qadeer A, Ahmad E, Zaman M, et al. Arch. Biochem. Biophys. 540(1-2), 101-16, (2013)

Loading...


Influences of perfluorooctanoic acid on the aggregation of multi-walled carbon nanotubes. Li C, Schäffer A, Vereecken H, et al. J. Environ. Sci. (China) 25(3), 466-72, (2013)

Loading...


Amphiphilic suramin dissolves Matrigel, causing an 'inhibition' artefact within in vitro angiogenesis assays. Prigozhina NL, Heisel AJ, Seldeen JR, et al. Int. J. Exp. Pathol. 94(6), 412-7, (2013)

Loading...


Biodegradation of anionic surfactant, sodium dodecyl sulphate by Pseudomonas aeruginosa MTCC 10311. Ambily PS and Jisha MS J. Environ Biol. 33(4), 717-20, (2012)

Loading...


Sodium dodecyl sulphate, a strong inducer of thermostable glucanhydrolase secretion from a derepressed mutant strain of Bacillus alcalophilus GCBNA-4. Shamim N, Ali S, and Ul-Haq I Appl. Biochem. Biotechnol. 169(8), 2467-77, (2013)

Loading...


Rgn gene is required for gut cell homeostasis after ingestion of sodium dodecyl sulfate in Drosophila. Pan J and Jin LH Gene 549(1), 141-8, (2014)

Loading...


Nanofiltration and Fenton's process over iron shavings for surfactants removal. Martins RC, Nunes M, Gando-Ferreira LM, et al. Environ. Technol. 35(17-20), 2380-8, (2014)

Loading...


Effect of anionic-nonionic-mixed surfactant micelles on solubilization of PAHs. Shi Z, Chen J, and Yin X J. Air Waste Manag. Assoc. 63(6), 694-701, (2013)

Loading...


Effects of sodium dodecyl sulphate on enhancement of lipoxygenase activity of hemoglobin. Ezebuo FC, Eze SO, and Chilaka FC Indian J. Exp. Biol. 50(12), 847-52, (2012)

Loading...


Effect of essential oils, hydrating agents, and ethanol on hair removal efficiency of thioglycolates. Moghimi HR, Jamali B, Farahmand S, et al. J. Cosmet. Dermatol. 12(1), 41-8, (2013)

Loading...


A non-destructive genotyping system from a single seed for marker-assisted selection in watermelon. Meru G, McDowell D, Waters V, et al. Genet. Mol. Res. 12(1), 702-9, (2013)

Loading...


Tolerance of droplet-digital PCR vs real-time quantitative PCR to inhibitory substances. Dingle TC, Sedlak RH, Cook L, et al. Clin. Chem. 59(11), 1670-2, (2013)

Loading...


eds. L.A. Lewis, J.J. Oppelt CRC Handbook of Electrophoresis Boca Raton, Florida 1, (1980)

Merck 14,8636

Beil. 1,IV,1847

Aldrich MSDS 1, 847:C / Corp MSDS 1 (1), 1495:C / FT-IR 2 (1), 1532:B / FT-IR 1 (1), 904:C / IR-Spectra (3), 544:C / IR-Spectra (2), 481:G / NMR-Reference 2 (2), 805:A / RegBook 1 (1), 1079:E / Sax 6, 2490 / Sigma FT-IR 1 (2), 70:B / Sigma FT-IR 1 (2), 70:C / Structure Index 1, 167:C:8

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?