71804 Fluka

Sodium oxalate

certified reference material (as reductometric standard), certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: Ethanedioic acid sodium salt, Oxalic acid disodium salt

  • CAS Number 62-76-0

  • Linear Formula NaOOCCOONa

  • Molecular Weight 134.00

  •  Beilstein Registry Number 3631622

  •  EC Number 200-550-3

  •  MDL number MFCD00012465

  •  PubChem Substance ID 24886305

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   certified reference material (as reductometric standard)
assay   ≥99.5%
quality   certified by BAM, according to ISO 17025

Description

Analysis Note

Exact concentration, expiry date, traceability and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Price and Availability

Customers Also Viewed

Sodium oxalate

BioXtra

Sodium oxalate

BioUltra, ≥99.5% (RT)

Sodium oxalate

puriss. p.a., ACS reagent, ≥99.5% (RT)

Sodium oxalate

≥99.99% trace metals basis

Sodium oxalate

ACS reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?