71943 Fluka

Sodium salicylate

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: 2-Hydroxybenzoic acid sodium salt, Natrii salicylas, Salicylic acid sodium salt

  • CAS Number 54-21-7

  • Linear Formula HOC6H4COONa

  • Molecular Weight 160.10

  •  Beilstein Registry Number 3732792

  •  EC Number 200-198-0

  •  MDL number MFCD00002440

  • Popular Documents:  RAMAN FTIR (PDF)  

Properties

Related Categories Pharmacopoeia, Pharmacopoeia A-Z More...
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia
mp   >300 °C(lit.)

Price and Availability

Customers Also Viewed

Sodium salicylate

ReagentPlus®, ≥99.5%

Sodium salicylate

meets USP testing specifications

Sodium salicylate

puriss. p.a., ≥99.5% (NT)

Sodium salicylate

puriss. p.a., Reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?