• USA Home
  • 72033 - Hydrochloric acid solution

EMAIL THIS PAGE TO A FRIEND
72033 Fluka

Hydrochloric acid solution

volumetric, 6 M HCl (6N)

Synonym: Hydrogen chloride solution

  • CAS Number 7647-01-0

  • Empirical Formula (Hill Notation) HCl

  • Molecular Weight 36.46

  •  MDL number MFCD00011324

Purchase

Properties

Related Categories Acid Solutions, Analytical Reagents, Analytical/Chromatography, Hydrochloric acid solution, Most common compounds for volumetric titration,
vapor pressure   190 mmHg ( 25 °C)
quality   volumetric
shelf life   limited shelf life
calibrated against   tris
concentration   6 M HCl (6N)
bp   100 °C(lit.)

Description

General description

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1789 8 / PGII
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?