72079 Fluka

Sodium hydroxide solution

~0.1 M NaOH, for HPCE

DOWNLOAD MSDS (PDF)
  • CAS Number 1310-73-2

  • Linear Formula NaOH

  • Molecular Weight 40.00

  •  EC Number 231-791-2

  •  MDL number MFCD00003548

  •  PubChem Substance ID 24886361

Properties

Related Categories Capillary Electrophoresis, Capillary Isoelectric Focusing, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Protein Electrophoresis,
grade   for HPCE
vapor pressure   3 mmHg ( 37 °C)
concentration   ~0.1 M NaOH
conductivity   ~17.5 mS/cm at 20 °C
abs.   UV absorption/200-800 nm in accordance
fluorescence   λex 200-600 nm; λem 300-800 nm (corresponds)

Description

Preparation Note

prepared under clean room conditions and filtered through a 0.2 μm membrane filter

Price and Availability

Customers Also Viewed

Hydrochloric acid solution

~0.1 M HCl, for HPCE

Sodium hydroxide concentrate

0.1 M NaOH in H2O (0.1N), Eluent concentrate for IC

Sodium hydroxide concentrate

for 1 L standard solution, 0.1 M NaOH (0.1 N)

Sodium hydroxide solution

~1.0 M NaOH, for HPCE

Sodium hydroxide solution

volumetric, 0.1 M NaOH (0.1N)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?