Synonym: 3′,4′,5,7-Tetrahydroxyflavone
CAS Number 491-70-3
Empirical Formula (Hill Notation) C15H10O6
Molecular Weight 286.24
Beilstein Registry Number 292084
EC Number 207-741-0
MDL number MFCD00017309
| Related Categories | Achillea, Alphabetical, Analytical Standards, Analytical/Chromatography, Chamomilla, |
| grade | analytical standard |
| assay | ≥97.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | ~330 °C(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?