• USA Home
  • 73426 - Nitrate Reduction Test

EMAIL THIS PAGE TO A FRIEND

73426 Fluka

Nitrate Reduction Test

for microbiology

Purchase

Properties

Related Categories Identification Tests & Reagents, Microbiology, Test Kits More...
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
storage temp.   2-8°C

Description

Components

Nitrate broth (Fluka 72548) 100 g
Sulfanilic acid (Fluka 86090) = Reagent A 100 g,
N,N-Dimethyl-1-naphthylamine (Sial D4011) = Reagent B 10 mL
Zinc (Fluka 93027) 1 g.

General description

Bacterial species may be differentiated on the basis of their ability to reduce nitrate to nitrite or nitrogenous gases. The reduction of nitrate may be coupled to anaerobic respiration in some species.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37-61
RIDADR 
UN 3316 9

Documents

Certificate of Analysis

Protocols & Articles

Articles

Differentiation of Escherichia coli from coliforms

In August 2008, the discovery of E. coli-contaminated beef in the United States prompted a nationwide recall of beef. The source turned out to be one supplier that had a history of contamination of i...
Jvo Siegrist
AnalytiX Volume 8 Article 5
Keywords: Decarboxylations, Fermentation, Genetic, Indicators, Microbiology, Reductions

Identification of Microorganisms Based on Color

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram. The Gram Stain allows the determination of morphology, dividing bacteria into tw...
Jvo Siegrist
Microbiology Focus Edition 1.4, Product Manager Microbiology.... ivo.siegrist@sial.com
Keywords: Antibiotics, Enzyme activity, Fermentation, Gram stain, Help, Indicators, Reductions

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?