Synonym: Blanc disks, Disks, sterile, Plain disks
| Related Categories | Discs and Strips, Identification Tests & Reagents, Microbiology More... |
| grade | for microbiology |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Can be used to test a variety of antibiotics, carbohydrates, substrates, antiseptics on bacteria in petri dishes. Soak a disc in a solution or apply some solution on the discs. Then let dry and place it in an inoculated agar plate. Each disc will absorb exactly the same amount of liquid.
Sterile filter paper discs (25 pcs) of 10 mm diameter
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?