74202 Fluka

Tetramethylammonium chloride

for ion pair chromatography, ≥99.0% (AT)

DOWNLOAD MSDS (PDF)

Synonym: TMA

  • CAS Number 75-57-0

  • Linear Formula (CH3)4N(Cl)

  • Molecular Weight 109.60

  •  Beilstein Registry Number 3911201

  •  EC Number 200-880-8

  •  MDL number MFCD00011628

  •  PubChem Substance ID 24886658

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Analytical/Chromatography, Buffers & Reagents, HPLC, Ion Pair Reagents, Ion Pair Reagents - Anionic More...
grade   for ion pair chromatography
assay   ≥99.0% (AT)
mp   >300 °C(lit.)
λ   10 % in H2O
UV absorption   λ: 220 nm Amax: 0.05
  λ: 230 nm Amax: 0.04
  λ: 250 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



NEW! Ascentis® Express 5 µm
HPLC Columns


New Ascentis Express 5 µm HPLC Columns!Ascentis Express columns, based on 5 µm Fused-Core® particles, offer better performance, faster and more efficient separations, compared to columns based on 3 µm and 5 µm totally porous particles, with no backpressure concerns!


Customers Also Viewed

Obidoxime chloride

≥95.0% (HPLC)

Tetramethylammonium chloride

BioUltra, for molecular biology, ≥99.0% (AT)

Tetramethylammonium chloride

purum, ≥98.0% (AT)

Tetramethylammonium chloride

reagent grade, ≥98%

Tetramethylammonium chloride solution

for molecular biology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?